missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl DL-Mandelate, TCI America™

Manufacturer:  TCI America M071125G

 View more versions of this product

Catalog No. M071125G



Specifications

Specifications

Benzyl DL-Mandelate
95°C
890-98-2
MFCD00021856
JFKWZVQEMSKSBU-UHFFFAOYSA-N
benzyl 2-hydroxy-2-phenylacetate
13461
Crystal-Powder at 20°C
White
25g
C15H14O3
DL-Mandelic Acid Benzyl Ester
C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O
242.274
242.27
≥98.0% (GC)

Chemical Identifiers

890-98-2
242.274
JFKWZVQEMSKSBU-UHFFFAOYSA-N
13461
C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O
C15H14O3
MFCD00021856
DL-Mandelic Acid Benzyl Ester
benzyl 2-hydroxy-2-phenylacetate
Safety and Handling

Safety and Handling

TSCA : No

Documents