missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl DL-Mandelate, TCI America™

$120.70 - $1,026.69

Chemical Identifiers

CAS 890-98-2
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD00021856
InChI Key JFKWZVQEMSKSBU-UHFFFAOYSA-N
Synonym DL-Mandelic Acid Benzyl Ester
PubChem CID 13461
IUPAC Name benzyl 2-hydroxy-2-phenylacetate
SMILES C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M071125G View Documents TCI America
M071125G
25g Each for $120.70
 
M0711500G View Documents TCI America
M0711500G
500g Each for $1,026.69
 
Specifications

Chemical Identifiers

890-98-2
242.274
JFKWZVQEMSKSBU-UHFFFAOYSA-N
13461
C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O
C15H14O3
MFCD00021856
DL-Mandelic Acid Benzyl Ester
benzyl 2-hydroxy-2-phenylacetate

Specifications

White
25g
C15H14O3
DL-Mandelic Acid Benzyl Ester
C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O
242.274
242.27
≥98.0% (GC)
95°C
890-98-2
MFCD00021856
JFKWZVQEMSKSBU-UHFFFAOYSA-N
benzyl 2-hydroxy-2-phenylacetate
13461
Crystal-Powder at 20°C
Benzyl DL-Mandelate
Safety and Handling

Safety and Handling

TSCA : No