Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl cinnamate, 99%

$48.22 - $164.24

Chemical Identifiers

Chemical Name or Material Benzyl cinnamate
CAS 103-41-3
Molecular Formula C16H14O2
Formula Weight 238.29
MDL Number MFCD00004789
Beilstein 2051339
Merck Index 14,1130
Synonym Cinnamic acid benzyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1955022

  SDS
alfa aesar™
A1955022
100g Each for $48.22

AAA1955036

  SDS
alfa aesar™
A1955036
500g Each for $164.24
Description & Specifications

Specifications

Formula Weight 238.29
Quantity 100g
RTECS Number GD8400000
Boiling Point 195° to 200°C (5mmHg)
CAS 103-41-3
TSCA TSCA
Molecular Formula C16H14O2
Merck Index 14,1130
Melting Point 34° to 36°C
Flash Point 180°C (356°F)
MDL Number MFCD00004789
Beilstein 2051339
Synonym Cinnamic acid benzyl ester
Assay 99%
Chemical Name or Material Benzyl cinnamate