missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyl cinnamate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1955022
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Benzyl cinnamate | |
34°C to 36°C | |
195°C to 200°C (5 mmHg) | |
C16H14O2 | |
100 g | |
14,1130 | |
NGHOLYJTSCBCGC-VAWYXSNFSA-N | |
benzyl (Z)-3-phenylprop-2-enoate | |
15558051 | |
99% |
103-41-3 | |
1.11 | |
180°C (356°F) | |
MFCD00004789,MFCD00004789,MFCD00004789 | |
2051339 | |
benzyl alcohol cinnamic ester, benzylcinnamate, benzyl alcohol, cinnamate, wln: r1u1vo1r, benzyl .gamma.-phenylacrylate, benzyl cis-cinnamate, benzyl z-cinnamate, 2-propenoic acid, 3-phenyl-, phenylmethyl ester, z-benzyl 3-phenylacrylate, cinnamic acid, benzyl ester, z | |
O=C(OCC1=CC=CC=C1)\C=C\C1=CC=CC=C1 | |
238.29 | |
238.29 |
Chemical Identifiers
103-41-3 | |
238.29 | |
NGHOLYJTSCBCGC-VAWYXSNFSA-N | |
15558051 | |
O=C(OCC1=CC=CC=C1)\C=C\C1=CC=CC=C1 |
C16H14O2 | |
MFCD00004789,MFCD00004789,MFCD00004789 | |
benzyl alcohol cinnamic ester, benzylcinnamate, benzyl alcohol, cinnamate, wln: r1u1vo1r, benzyl .gamma.-phenylacrylate, benzyl cis-cinnamate, benzyl z-cinnamate, 2-propenoic acid, 3-phenyl-, phenylmethyl ester, z-benzyl 3-phenylacrylate, cinnamic acid, benzyl ester, z | |
benzyl (Z)-3-phenylprop-2-enoate |
Safety and Handling
EINECSNumber : 203-109-3
RTECSNumber : GD8400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only