Learn More
Benzyl chloromethyl ether, 75%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 376952500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Benzyl chloromethyl ether | |
70.0 | |
1.1300g/mL | |
91°C | |
70% min. (GC) | |
C8H9ClO | |
MFCD00000886 | |
1.13 | |
LADPCMZCENPFGV-UHFFFAOYSA-N | |
chloromethoxymethylbenzene | |
137983 | |
75% |
3587-60-8 | |
100.0 | |
102°C (14.0mmHg) | |
Authentic | |
Glass bottle | |
C6H5CH2OCH2Cl | |
250g | |
benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl | |
C1=CC=C(C=C1)COCCl | |
156.61 | |
156.61 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause cancer.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handl
GHS Signal Word: Danger
RUO â Research Use Only