Learn More
Benzyl chloroformate, 97 wt%, stabilized, Thermo Scientific Chemicals
Benzyl chloroformate, 97 wt%, stabilized, Acros Organics, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 178.0 deg.C to 180.0 deg.C, Undesignated, Melting Point: -20.0 deg.C, Molecular Weight: 170.6, Percent Purity: 97 wt%, Assay Percent Range: 96% min. (GC), Beilstein: 06,437
Supplier: Thermo Scientific Chemicals 152945000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentSpecifications
Benzyl chloroformate | |
-20.0°C | |
178.0°C to 180.0°C | |
Authentic | |
Glass bottle | |
1.5160 to 1.5180 | |
500 g | |
01,109; 02,59; 15,22 | |
15,1801 | |
Solubility in water: reacts | |
C1=CC=C(C=C1)COC(=O)Cl | |
170.6 | |
10387 | |
170.6 | |
Liquid |
501-53-1,100-44-7 | |
1.2000g/mL | |
92°C | |
96% min. (GC) | |
C8H7ClO2 | |
ClCO2CH2C6H5 | |
06,437 | |
1.2 | |
benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester | |
HSDAJNMJOMSNEV-UHFFFAOYSA-N | |
benzyl carbonochloridate | |
0.1% anhydrous sodium carbonate | |
2.36 mPa.s (20°C) | |
97 wt% |
Chemical Identifiers
501-53-1 | |
170.6 | |
benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester | |
benzyl carbonochloridate |
C8H7ClO2 | |
HSDAJNMJOMSNEV-UHFFFAOYSA-N | |
10387 | |
C1=CC=C(C=C1)COC(=O)Cl |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Harmful if inhaled.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 207-925-
Recommended Storage : Important Note: the material builds progressively benzyl chloride under normal storage conditions
RUO – Research Use Only