missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl chloroformate, 95%, stab. with ca 0.1% sodium carbonate

Manufacturer:  Alfa Aesar™ A1568230

 View more versions of this product

Catalog No. AAA1568230



Description

Description

Applications
Benzyl chloroformate is widely used as a reactive chemical intermediate in plastic, pharmaceutical, agricultural and organic chemicals. It is useful for the introduction of carboxybenzyl (cbz) protecting group for amines such as aniline in organic synthesis. It is also involved in the synthesis of 1,2,4-oxadiazoles.

Solubility
Miscible with ether, acetone and benzene.

Notes
Store in cool place. Moisture sensitive.
Specifications

Specifications

Benzyl chloroformate
Acrid
-20°C
stabilized with ≈0.1% sodium carbonate
C8H7ClO2
UN1739
benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
C1=CC=C(C=C1)COC(=O)Cl
170.592
170.6
125°C (257°F)
178°C to 180°C
250g
501-53-1
MFCD00000640
14,1801
HSDAJNMJOMSNEV-UHFFFAOYSA-N
benzyl carbonochloridate
10387
95%

Chemical Identifiers

501-53-1
170.592
HSDAJNMJOMSNEV-UHFFFAOYSA-N
10387
C1=CC=C(C=C1)COC(=O)Cl
C8H7ClO2
MFCD00000640
benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
benzyl carbonochloridate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P273-P305+P351+P338-P309-P310-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: BENZYL CHLOROFORMATE

TSCA : TSCA

Recommended Storage : −30° to −10°C

Documents