Learn More
Benzyl chloroformate, 3M (50 wt%) solution in toluene, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 124495000
Description
Coupling reagentSpecifications
Benzyl chloroformate | |
7°C | |
1.01 | |
500mL | |
501-53-1,108-88-3,100-44-7 | |
53.0,55.0 | |
C6H5CH2OOCCl | |
06, 437 | |
15, 1801 | |
Solubility in water: decomposes | |
C1=CC=C(C=C1)COC(=O)Cl | |
170.6 | |
170.6 | |
45 to 53 wt% |
1.0100g/mL | |
Glass bottle | |
Pink to Yellow | |
50 wt. % Solution in Toluene | |
45.0,47.0 | |
C8H7ClO2 | |
MFCD00000640 | |
01,109; 02,59; 15,22 | |
benzyl chlorocarbonate, benzyl chloroformate, benzylcarbonyl chloride, benzyloxycarbonyl chloride, carbobenzoxy chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, cbz chloride, cbz-cl, chloroformic acid, benzyl ester | |
HSDAJNMJOMSNEV-UHFFFAOYSA-N | |
benzyl carbonochloridate | |
10387 | |
Solution |
Chemical Identifiers
501-53-1 | |
170.6 | |
HSDAJNMJOMSNEV-UHFFFAOYSA-N | |
10387 | |
C1=CC=C(C=C1)COC(=O)Cl |
C8H7ClO2 | |
MFCD00000640 | |
benzyl chlorocarbonate, benzyl chloroformate, benzylcarbonyl chloride, benzyloxycarbonyl chloride, carbobenzoxy chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, cbz chloride, cbz-cl, chloroformic acid, benzyl ester | |
benzyl carbonochloridate |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness. May cause cancer. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. Suspected of damaging the unborn child. Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Avoid release to the environment.
GHS Signal Word: Danger
EINECSNumber : 207-925-0
TSCA : TSCA