Please login to your online account to display your discounted pricing

Benzyl chloride, 99%, ACROS Organics™


Quantity 250g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  405092500

 View more versions of this product

Catalog No. AC405092500

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Benzyl chloride
Name Note Stabilized, Pure
CAS 100-44-7
CAS Min % 98.5
Health Hazard 1 Danger
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Fatal if inhaled.
May cause cancer.
May cause respiratory irritation.
Causes skin irritation.
May cause genetic defects.
May cause damage to organs through prolonged or repeated exposure.
CAS 75-56-9
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.5370 to 1.5390 (20°C, 589nm)
Stabilizer 0.25% propylene oxide
Purity Grade Pure
Color Clear colorless to slightly yellow
Physical Form Liquid
Assay ≥98.5%
Assay Percent Range 99%
Quantity 250g
Molecular Formula C7H7Cl
Linear Formula C6H5CH2Cl
Formula Weight 126.59
MDL Number MFCD00000889

Specifications

Chemical Name or Material Benzyl chloride
Name Note Stabilized, Pure
CAS 100-44-7
CAS Min % 98.5
CAS Max % 100.0
CAS 75-56-9
Purity Grade Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H7Cl
Linear Formula C6H5CH2Cl
Formula Weight 126.59
MDL Number MFCD00000889
Accessories

Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Fatal if inhaled.
May cause cancer.
May cause respiratory irritation.
Causes skin irritation.
May cause genetic defects.
May cause damage to organs through prolonged or repeated exposure.

SDS & Safety Documents