Please login to your online account to display your discounted pricing

Benzyl chloride, 99.5+%, extra pure, stabilized, ACROS Organics™


Quantity 2.5L
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  180850025

 View more versions of this product

Catalog No. AC180850025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.50%
Purity Grade Extra Pure
Physical Form Liquid
Formula Weight 126.59
Quantity 2.5L
Color Clear
Packaging Glass Bottle
EINECS Number 202-853-6
TSCA TSCA
RTECS Number XS8925000
CAS Max % 100.0
Refractive Index 1.5370 to 1.5390 (20°C, 589nm)
Name Note 99.5+%
Assay Percent Range 99.5+%
MDL Number MFCD00000889
CAS 100-44-7
Health Hazard 1 Danger
CAS 75-56-9
CAS Min % 99.5
Chemical Name or Material Benzyl chloride
Linear Formula C6H5CH2Cl
Molecular Formula C7H7Cl
Infrared Spectrum Authentic
Stabilizer 0.25% propylene oxide
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Fatal if inhaled.
May cause cancer.
May cause respiratory irritation.
Causes skin irritation.
May cause genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Benzyl chloride
Name Note 99.5+%
CAS 100-44-7
CAS Min % 99.5
CAS Max % 100.0
CAS 75-56-9
Purity Grade Extra Pure
Assay ≥99.50%
Assay Percent Range 99.5+%
Molecular Formula C7H7Cl
Linear Formula C6H5CH2Cl
Formula Weight 126.59
MDL Number MFCD00000889
Accessories

Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Fatal if inhaled.
May cause cancer.
May cause respiratory irritation.
Causes skin irritation.
May cause genetic defects.
May cause damage to organs through prolonged or repeated exposure.

SDS & Safety Documents