missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl chloride, 99.5+%, extra pure, stabilized, ACROS Organics™

Chemical Identifiers

CAS 100-44-7
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.59
MDL Number MFCD00000889
InChI Key KCXMKQUNVWSEMD-UHFFFAOYSA-N
Synonym alpha-chlorotoluene, alpha-chlortoluol, benzene, chloromethyl, benzyl chloride, benzylchlorid, benzylchloride, chloromethyl benzene, chlorophenylmethane, chlorure de benzyle, tolyl chloride
PubChem CID 7503
ChEBI CHEBI:615597
IUPAC Name chloromethylbenzene
SMILES C1=CC=C(C=C1)CCl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC180850100
SDS View Documents
Thermo Fisher Scientific
AC180850100
10L Plastic drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Alkylation reagent
Specifications

Chemical Identifiers

100-44-7
126.59
KCXMKQUNVWSEMD-UHFFFAOYSA-N
7503
chloromethylbenzene
C7H7Cl
MFCD00000889
alpha-chlorotoluene, alpha-chlortoluol, benzene, chloromethyl, benzyl chloride, benzylchlorid, benzylchloride, chloromethyl benzene, chlorophenylmethane, chlorure de benzyle, tolyl chloride
CHEBI:615597
C1=CC=C(C=C1)CCl

Specifications

1.1000g/mL
Authentic
1.5370 to 1.5390
0.25% propylene oxide
179.0°C
-39.0°C
100-44-7,75-56-9
100.0
C7H7Cl
MFCD00000889
15,1132
Solubility in water: 0.3g/L (20°C). Other solubilities: miscible with ethanol,ethyl ether,chloroform,,acetone,slightly soluble in carbon tetrachloride
C1=CC=C(C=C1)CCl
126.59
CHEBI:615597
Liquid
Extra Pure
67°C
Plastic drum
1.1
1.280 mPa.s (20°C)
Undesignated
10L
99.5
99.5% min. (GC)
C6H5CH2Cl
05,292
alpha-chlorotoluene, alpha-chlortoluol, benzene, chloromethyl, benzyl chloride, benzylchlorid, benzylchloride, chloromethyl benzene, chlorophenylmethane, chlorure de benzyle, tolyl chloride
KCXMKQUNVWSEMD-UHFFFAOYSA-N
chloromethylbenzene
7503
126.59
99.5+%
Benzyl chloride, 99.5+%
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Toxic if inhaled.
May cause cancer.
May cause respiratory irritation.
Causes skin irritation.
May cause genetic defects.
May cause damage to organs through prolonged or

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautio

GHS Signal Word: Danger

EINECSNumber : 202-853-6

RTECSNumber : XS8925000

TSCA : TSCA

SDS