missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyl carbamate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1156914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
85°C to 89°C | |
621-84-1 | |
MFCD00007965 | |
carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421 | |
NC(=O)OCC1=CC=CC=C1 | |
151.17 | |
151.17 | |
25 g |
Benzyl carbamate | |
C8H9NO2 | |
1865635 | |
PUJDIJCNWFYVJX-UHFFFAOYSA-N | |
benzyl carbamate | |
12136 | |
99% |
Chemical Identifiers
621-84-1 | |
151.17 | |
PUJDIJCNWFYVJX-UHFFFAOYSA-N | |
12136 | |
NC(=O)OCC1=CC=CC=C1 |
C8H9NO2 | |
MFCD00007965 | |
carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421 | |
benzyl carbamate |
Safety and Handling
P261-P271-P304+P340-P312-P501c
H332-H335
EINECSNumber : 210-710-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only