Please login to your online account to display your discounted pricing

Benzyl butyl phthalate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Benzyl butyl phthalate
CAS 85-68-7
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C19H20O4
Formula Weight 312.37g/mol
Synonym BBP
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC430170250

  SDS
thermo fisher scientific
430170250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5390 to 1.5410 (20°C, 589nm)
Density 1.1g/mL
Color Clear colorless
Quantity 25mL
Chemical Name or Material Benzyl butyl phthalate
CAS 85-68-7
CAS Min % 96.0
CAS Max % 100.0
Physical Form Clear colorless viscous liquid
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C19H20O4
Formula Weight 312.37g/mol
Synonym BBP
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May damage the unborn child. Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May damage the unborn child. Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.