missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl Bromide, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1353518*

 View more versions of this product

Catalog No. AAA1353518



Description

Description

Applications
Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographic chemicals, synthetic resins, dyes and plasticizers and esters. It is also involved in the synthesis of (E)-(2-Iodovinyl)benzene by reacting it with diiodomethane.

Solubility
Miscible with benzene, carbon tetrachloride, ethanol and ether.

Notes
Light and moisture sensitive. Incompatible with bases, alcohols, amines, oxidizing agents and corrodes steel.
Specifications

Specifications

Benzyl Bromide
87°C
-4°C
100-39-0
C7H7Br
AGEZXYOZHKGVCM-UHFFFAOYSA-N
bromomethylbenzene
7498
171.04
1.438g/mL
198°C to 199°C
50g
99%
benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
C1=CC=C(C=C1)CBr
171.037
CHEBI:59858
Liquid

Chemical Identifiers

100-39-0
171.037
benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
CHEBI:59858
C1=CC=C(C=C1)CBr
C7H7Br
AGEZXYOZHKGVCM-UHFFFAOYSA-N
7498
bromomethylbenzene
Safety and Handling

Safety and Handling

storageNote1 : Moisture sensitive

SDS Documents