Please login to your online account to display your discounted pricing

Benzyl bromide, 99%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Benzyl bromide
CAS 100-39-0
Molecular Formula C7H7Br
Formula Weight 171.04
MDL Number MFCD00000172
UN Number UN1737
Beilstein 385801
Merck Index 14,1128
Synonym alpha-Bromotoluene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA135350B

  SDS
alfa aesar
A135350B
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.04
Quantity 1000g
TSCA TSCA
Refractive Index 1.5750
Density 1.438
Melting Point −4°C
Boiling Point 198° to 199°C
Flash Point 87°C(189°F)
Molecular Formula C7H7Br
Odor Pungent, Makes Eyes Water
MDL Number MFCD00000172
CAS 100-39-0
UN Number UN1737
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H335-H227-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material Benzyl bromide
Physical Form Liquid
Assay 99%
Sensitivity Moisture Sensitive
Beilstein 385801
Merck Index 14,1128
Synonym alpha-Bromotoluene
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BENZYL BROMIDE
RTECS Number XS7965000

Applications
Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographic chemicals, synthetic resins, dyes and plasticizers and esters. It is also involved in the synthesis of (E)-(2-Iodovinyl)benzene by reacting it with diiodomethane.

Solubility
Miscible with benzene, carbon tetrachloride, ethanol and ether.

Notes
Light and moisture sensitive. Incompatible with bases, alcohols, amines, oxidizing agents and corrodes steel.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H335-H227-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes serious eye irritation.