missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl azide, 94%, Alfa Aesar

Manufacturer:  Alfa Aesar 3263509

 View more versions of this product

Catalog No. AA3263509



Specifications

Specifications

Benzyl Azide
43°C
10g
94%
azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029, benzene, azidomethyl
C1=CC=C(C=C1)CN=[N+]=[N-]
133.154
133.15
1.0655g/mL
82 to 85° (16mm)
622-79-7
C7H7N3
UDLLFLQFQMACJB-UHFFFAOYSA-N
azidomethylbenzene
12152
Liquid

Chemical Identifiers

622-79-7
133.154
azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029, benzene, azidomethyl
azidomethylbenzene
C7H7N3
UDLLFLQFQMACJB-UHFFFAOYSA-N
12152
C1=CC=C(C=C1)CN=[N+]=[N-]
SDS Documents