Please login to your online account to display your discounted pricing

Benzyl azide, 94%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Benzyl azide
CAS 622-79-7
Molecular Formula C6H5CH2N3
Formula Weight 133.15
MDL Number MFCD00013836
UN Number UN1993
Synonym Triazotoluene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3263518

  SDS
alfa aesar
3263518
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 133.15
Quantity 50g
Chemical Name or Material Benzyl azide
Physical Form Liquid
Assay 94%
Molecular Formula C6H5CH2N3
MDL Number MFCD00013836
CAS 622-79-7
UN Number UN1993
Synonym Triazotoluene
Solubility Information Insoluble in water
Health Hazard 1 Warning
Refractive Index 1.5341
Health Hazard 2 GHS H Statement
H226;
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.065
Boiling Point 82° to 85°C (16mmHg)
Color Colorless to Pale Brown
Flash Point 43°C (110°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECS Number XS6650000
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226;
Flammable liquid and vapour.