missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl Alcohol 99.0+%, TCI America™

Manufacturer:  Tci America B2378500G

Catalog No. B2378500G



Specifications

Specifications

Benzyl Alcohol
Yellow
500g
C7H8O
alpha-toluenol
C1=CC=C(C=C1)CO
108.14
CHEBI:17987
Clear Liquid at 20°C
200°C
-15°C
100-51-6
MFCD00004599
WVDDGKGOMKODPV-UHFFFAOYSA-N
phenylmethanol
244
108.14
≥99.0% (GC)

Chemical Identifiers

100-51-6
108.14
WVDDGKGOMKODPV-UHFFFAOYSA-N
244
phenylmethanol
C7H8O
MFCD00004599
alpha-toluenol
CHEBI:17987
C1=CC=C(C=C1)CO
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1011

RTECSNumber : DN3150000

TSCA : Yes

Documents