Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl alcohol on polystyrene, 3.5 mmol/g

$153.93 - $153.93

Chemical Identifiers

Chemical Name or Material Benzyl alcohol on polystyrene
MDL Number MFCD03792087
Synonym Poly(4-hydroxymethyl)styrene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1959206

 
alfa aesar™
L1959206
5g Each for $153.93

AAL1959214

 
alfa aesar™
L1959214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Benzyl alcohol on polystyrene
MDL Number MFCD03792087
Synonym Poly(4-hydroxymethyl)styrene
Concentration 3.5mmol/g
Quantity 5g