Please login to your online account to display your discounted pricing

Benzyl Alcohol NF, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Benzyl Alcohol
CAS 100-51-6
Purity Grade NF
Molecular Formula C6H5CH2OH
Formula Weight 108.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK14031

 
avantor performance
140312
Glass Bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK14034

 
avantor performance
140310*
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 108.14
Quantity 1L
Packaging Glass Bottle
Purity Grade NF
Chemical Name or Material Benzyl Alcohol
CAS 100-51-6
Molecular Formula C6H5CH2OH