missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Riedel-de Haen™ Benzyl alcohol, ACS Reagent, ≥99%, Honeywell™ Riedel-de Haën™

ACS Reagent,≥99.0%

$194.89 - $600.57

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES C1=CC=C(C=C1)CO
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046788 View Documents Honeywell Riedel-de Haen™
4028341L
1L Glass Bottle Each for $385.28
 
60046787 View Documents Honeywell Riedel-de Haen™
402834100ML
100mL Glass Bottle Each for $194.89
 
60046789 View Documents Honeywell Riedel-de Haen™
40283425L
2.5L Glass Bottle Each for $600.57
 
60046790 View Documents Honeywell Riedel-de Haen™
402834500ML
500mL Glass Bottle Each for $273.17
 
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

100-51-6
108.14
WVDDGKGOMKODPV-UHFFFAOYSA-N
244
phenylmethanol
C7H8O
MFCD00004599
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
CHEBI:17987
C1=CC=C(C=C1)CO

Specifications

1.045g/mL at 25°C
817°F
Glass Bottle
3.7 (vs air)
203°C to 205°C
-16°C to -13°C
100-51-6
C6H5CH2OH
UN3334
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
C1=CC=C(C=C1)CO
108.14
CHEBI:17987
Liquid
Benzyl alcohol
96°C (204.8°F)
≤0.005%
n20/D 1.539
13.3mmHg (100°C), 3.75mmHg (77°C)
Undesignated
1L
C7H8O
MFCD00004599
878307
WVDDGKGOMKODPV-UHFFFAOYSA-N
phenylmethanol
244
108.14g/mol
≥99%
Safety and Handling

Safety and Handling

P261-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338

DOTInformation : UN 3334

RTECSNumber : 202-859-9