Please login to your online account to display your discounted pricing

Benzyl alcohol, ACS, 99+%, Alfa Aesar™

$134.41 - $134.41

Chemical Identifiers

Chemical Name or Material Benzyl alcohol
CAS 100-51-6
Purity Grade ACS Grade
Molecular Formula C7H8O
Formula Weight 108.14
MDL Number MFCD00004599
Beilstein 878307
Merck Index 14,1124
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA41218K2

  SDS
alfa aesar
41218K2
Each for $134.41
Description & Specifications

Specifications

Chemical Name or Material Benzyl alcohol
CAS 100-51-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332;
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DN3150000
TSCA TSCA
Purity Grade ACS Grade
Physical Form Liquid
Assay ≥99%
Molecular Formula C7H8O
Formula Weight 108.14
MDL Number MFCD00004599
Beilstein 878307
Merck Index 14,1124
Refractive Index 1.5400
Density 1.045
Boiling Point 205°C
Melting Point −15°C
Quantity 1L
Flash Point 100°C (212°F)
Odor Aromatic
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332;
Harmful if swallowed.
Harmful if inhaled.