Learn More
Benzyl alcohol, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 447721000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Benzyl alcohol | |
-15.0°C | |
205.0°C | |
Glass Bottle | |
1.5380 to 1.5410 | |
MFCD00004599,MFCD03792087 | |
1.044 | |
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
WVDDGKGOMKODPV-UHFFFAOYSA-N | |
phenylmethanol | |
244 | |
CHEBI:17987 | |
ACS Reagent |
100-51-6 | |
1.0440g/mL | |
96°C | |
C7H8O | |
C6H5CH2OH | |
100 mL | |
15,1127 | |
Solubility in water: 40g/L (20°C). Other solubilities: miscible with alcohol,ether,acetone,chloroform,,dmf and methylene chloride | |
OCC1=CC=CC=C1 | |
108.14 | |
6.6 mPa.s (20°C) | |
108.14 |
Chemical Identifiers
100-51-6 | |
108.14 | |
WVDDGKGOMKODPV-UHFFFAOYSA-N | |
244 | |
phenylmethanol |
C7H8O | |
MFCD00004599,MFCD03792087 | |
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
CHEBI:17987 | |
OCC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/phys
GHS Signal Word: Warning
EINECSNumber : 202-859-9
RUO – Research Use Only