Learn More
Benzyl alcohol, ACS reagent, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC447720025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Benzyl Alcohol | |
1.0440g/mL | |
205.0°C | |
C7H8O | |
MFCD00004599,MFCD03792087 | |
15,1127 | |
alpha-toluenol, benzenecarbinol, benzenemethanol, benzoyl alcohol, benzyl alcohol, benzylalcohol, hydroxymethyl benzene, hydroxytoluene, phenylcarbinol, phenylmethyl alcohol | |
Solubility in water: 40g/L (20°C). Other solubilities: miscible with alcohol,ether,acetone,chloroform,,dmf and methylene chloride | |
1.044 | |
phenylmethanol | |
244 | |
108.14 | |
6.6 mPa.s (20°C) |
100-51-6 | |
-15.0°C | |
96°C | |
C6H5CH2OH | |
Glass Bottle | |
1.5380 to 1.5410 | |
2.5L | |
WVDDGKGOMKODPV-UHFFFAOYSA-N | |
OCC1=CC=CC=C1 | |
108.14 | |
CHEBI:17987 | |
ACS Reagent |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/phys
GHS Signal Word: Warning
EINECSNumber : 202-859-9
RUO – Research Use Only