missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyl alcohol, ACS reagent, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC447720010
Specifications
Benzyl Alcohol | |
96°C | |
1.5380 to 1.5410 | |
6.6 mPa.s (20°C) | |
-15.0°C | |
100-51-6 | |
C6H5CH2OH | |
alpha-toluenol, benzenecarbinol, benzenemethanol, benzoyl alcohol, benzyl alcohol, benzylalcohol, hydroxymethyl benzene, hydroxytoluene, phenylcarbinol, phenylmethyl alcohol | |
WVDDGKGOMKODPV-UHFFFAOYSA-N | |
phenylmethanol | |
244 | |
108.14 |
1.0440g/mL | |
Glass Bottle | |
1.044 | |
205.0°C | |
1L | |
C7H8O | |
15,1127 | |
Solubility in water: 40g/L (20°C). Other solubilities: miscible with alcohol,ether,acetone,chloroform,,dmf and methylene chloride | |
C1=CC=C(C=C1)CO | |
108.14 | |
CHEBI:17987 | |
ACS Reagent |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/phys
GHS Signal Word: Warning
EINECSNumber : 202-859-9