missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl acetoacetate, 97%

Manufacturer:  Alfa Aesar™ A1491530

 View more versions of this product

Catalog No. AAA1491530



Description

Description

Applications
Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.

Solubility
It is soluble in alkali solutions.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

Benzyl acetoacetate
276°C
5396-89-4
MFCD00008785
benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x
CC(=O)CC(=O)OCC1=CC=CC=C1
192.214
192.22
108°C (226°F)
250g
C11H12O3
2049615
WOFAGNLBCJWEOE-UHFFFAOYSA-N
benzyl 3-oxobutanoate
142266
97%

Chemical Identifiers

5396-89-4
192.214
WOFAGNLBCJWEOE-UHFFFAOYSA-N
142266
CC(=O)CC(=O)OCC1=CC=CC=C1
C11H12O3
MFCD00008785
benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x
benzyl 3-oxobutanoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

SDS Documents