Learn More
Benzyl acetate, 99%, Thermo Scientific Chemicals
$72.33 - $214.67
Chemical Identifiers
CAS | 140-11-4 |
---|---|
Molecular Formula | C9H10O2 |
Molecular Weight (g/mol) | 150.177 |
MDL Number | MFCD00008712 |
InChI Key | QUKGYYKBILRGFE-UHFFFAOYSA-N |
Synonym | acetic acid benzyl ester, benzyl ethanoate, acetic acid, phenylmethyl ester, phenylmethyl acetate, acetic acid, benzyl ester, alpha-acetoxytoluene, phenylmethyl ethanoate, acetoxymethyl benzene, benzylester kyseliny octove, caswell no. 081ea |
PubChem CID | 8785 |
ChEBI | CHEBI:52051 |
IUPAC Name | benzyl acetate |
SMILES | CC(=O)OCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2347236
|
Thermo Scientific Chemicals
B2347236 |
500 g |
Each for $72.33
|
|
|||||
AAB234720E
|
Thermo Scientific Chemicals
B234720E |
2500 g |
Each for $214.67
|
|
|||||
Description
Benzyl acetate can be used as a high boiling point solvent in coatings, like ink coating, binding agent and paint remover. It is used in soap and other chemical essences and has the effect of promote in floral and fantasy essences in jasmine, white orchid, fragrant plantain lily, gekkakou, narcissus and other essences. It is a plasticizer for ionophore membranes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzyl acetate can be used as a high boiling point solvent in coatings, like ink coating, binding agent and paint remover. It is used in soap and other chemical essences and has the effect of promote in floral and fantasy essences in jasmine, white orchid, fragrant plantain lily, gekkakou, narcissus and other essences. It is a plasticizer for ionophore membranes.
Solubility
Soluble in water 3.1 g/L.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Chemical Identifiers
140-11-4 | |
150.177 | |
QUKGYYKBILRGFE-UHFFFAOYSA-N | |
8785 | |
benzyl acetate |
C9H10O2 | |
MFCD00008712 | |
acetic acid benzyl ester, benzyl ethanoate, acetic acid, phenylmethyl ester, phenylmethyl acetate, acetic acid, benzyl ester, alpha-acetoxytoluene, phenylmethyl ethanoate, acetoxymethyl benzene, benzylester kyseliny octove, caswell no. 081ea | |
CHEBI:52051 | |
CC(=O)OCC1=CC=CC=C1 |
Specifications
140-11-4 | |
1.053 | |
95°C (203°F) | |
C9H10O2 | |
MFCD00008712 | |
1908121 | |
acetic acid benzyl ester, benzyl ethanoate, acetic acid, phenylmethyl ester, phenylmethyl acetate, acetic acid, benzyl ester, alpha-acetoxytoluene, phenylmethyl ethanoate, acetoxymethyl benzene, benzylester kyseliny octove, caswell no. 081ea | |
QUKGYYKBILRGFE-UHFFFAOYSA-N | |
benzyl acetate | |
8785 | |
150.18 | |
Liquid |
°C to 51°C | |
205°C to 207°C | |
99+% | |
1.502 | |
500 g | |
14,1123 | |
Soluble in water 3.1g/L. | |
CC(=O)OCC1=CC=CC=C1 | |
150.177 | |
CHEBI:52051 | |
99% | |
Benzyl acetate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-399-7
RTECSNumber : AF5075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only