Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl 6-hydroxynicotinate, 99%

$82.50 - $826.59

Chemical Identifiers

Chemical Name or Material Benzyl 6-hydroxynicotinate
CAS 191157-01-4
Molecular Formula C13H11NO3
Formula Weight 229.24
MDL Number MFCD02683072
Synonym Benzyl 6-hydroxypyridine-3-carboxylate; 6-Hydroxynicotinic acid benzyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2180706

 
alfa aesar™
B2180706
5g Each for $82.50

AAB2180714

 
alfa aesar™
B2180714
25g Each for $294.00

AAB2180722

 
alfa aesar™
B2180722
100g Each for $826.59
Description & Specifications

Specifications

Formula Weight 229.24
Chemical Name or Material Benzyl 6-hydroxynicotinate
Assay 99%
Molecular Formula C13H11NO3
MDL Number MFCD02683072
CAS 191157-01-4
Synonym Benzyl 6-hydroxypyridine-3-carboxylate; 6-Hydroxynicotinic acid benzyl ester
Melting Point 178° to 181°C
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.