missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl 4-oxo-1-piperidinecarboxylate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC425960250

 View more versions of this product

Catalog No. AC425960250


 

Specifications Description & Specifications

Specifications

Solid or Lliquid
233.27
25mL
100.0
Glass bottle
19099-93-5
Benzyl 4-oxo-1-piperidinecarboxylate
Authentic
≥98.5%
White to pale yellow or clear
1.1850g/mL
96.0
C13H15NO3
97%
MFCD00673144

Chemical Identifiers

19099-93-5
100.0
C13H15NO3
MFCD00673144
≥98.5%
96.0
97%
233.27
Benzyl 4-oxo-1-piperidinecarboxylate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.