Please login to your online account to display your discounted pricing

Benzyl 4-oxo-1-piperidinecarboxylate, 97%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC425960050

 View more versions of this product

Catalog No. AC425960050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color White to pale yellow or clear
Formula Weight 233.27
Physical Form Solid or Lliquid
Assay ≥98.5%
Quantity 5mL
Density 1.1850g/mL
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
CAS 19099-93-5
Molecular Formula C13H15NO3
Chemical Name or Material Benzyl 4-oxo-1-piperidinecarboxylate
Assay Percent Range 97%
MDL Number MFCD00673144
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Benzyl 4-oxo-1-piperidinecarboxylate
CAS 19099-93-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 97%
Molecular Formula C13H15NO3
Formula Weight 233.27
MDL Number MFCD00673144
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents