Please login to your online account to display your discounted pricing

Benzyl 4-oxo-1-piperidinecarboxylate, 97%, ACROS Organics™

$199.92 - $199.92

Chemical Identifiers

Chemical Name or Material Benzyl 4-oxo-1-piperidinecarboxylate
CAS 19099-93-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 97%
Molecular Formula C13H15NO3
Formula Weight 233.27
MDL Number MFCD00673144
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC425960250

  SDS
thermo fisher scientific
AC425960250
Glass bottle 25mL Each for $199.92

AC425960050

  SDS
thermo fisher scientific
AC425960050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solid or Lliquid
Assay ≥98.5%
Formula Weight 233.27
Color White to pale yellow or clear
Quantity 25mL
Density 1.1850g/mL
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Molecular Formula C13H15NO3
CAS 19099-93-5
Assay Percent Range 97%
Chemical Name or Material Benzyl 4-oxo-1-piperidinecarboxylate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00673144
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.