missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl 4-Hydroxy-1-piperidinecarboxylate, TCI America™

$58.06 - $198.61

Chemical Identifiers

CAS 95798-23-5
Molecular Formula C13H17NO3
Molecular Weight (g/mol) 235.283
MDL Number MFCD01863722
InChI Key JKIUUDJOCYHIGY-UHFFFAOYSA-N
Synonym benzyl 4-hydroxy-1-piperidinecarboxylate, 1-cbz-4-hydroxypiperidine, n-cbz-4-hydroxy-1-piperidine, benzyl 4-hydroxytetrahydro-1 2h-pyridinecarboxylate, n-benzyloxycarbonyl-4-hydroxypiperidine, 1-piperidinecarboxylic acid, 4-hydroxy-, phenylmethyl ester, n-cbz-4-hydroxypiperidine, n-benzyloxycarbonyl piperidin-4-ol, 4-hydroxypiperidine, n-cbz protected, 4-hydroxy-piperidine-1-carboxylic acid benzyl ester
PubChem CID 2794620
IUPAC Name benzyl 4-hydroxypiperidine-1-carboxylate
SMILES C1CN(CCC1O)C(=O)OCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B48695G View Documents TCI America
B48695G
5g Each for $58.06
 
B486925G View Documents TCI America
B486925G
25g Each for $198.61
 
Specifications

Chemical Identifiers

95798-23-5
235.283
JKIUUDJOCYHIGY-UHFFFAOYSA-N
2794620
C1CN(CCC1O)C(=O)OCC2=CC=CC=C2
C13H17NO3
MFCD01863722
benzyl 4-hydroxy-1-piperidinecarboxylate, 1-cbz-4-hydroxypiperidine, n-cbz-4-hydroxy-1-piperidine, benzyl 4-hydroxytetrahydro-1 2h-pyridinecarboxylate, n-benzyloxycarbonyl-4-hydroxypiperidine, 1-piperidinecarboxylic acid, 4-hydroxy-, phenylmethyl ester, n-cbz-4-hydroxypiperidine, n-benzyloxycarbonyl piperidin-4-ol, 4-hydroxypiperidine, n-cbz protected, 4-hydroxy-piperidine-1-carboxylic acid benzyl ester
benzyl 4-hydroxypiperidine-1-carboxylate

Specifications

167°C
5g
C13H17NO3
benzyl 4-hydroxy-1-piperidinecarboxylate, 1-cbz-4-hydroxypiperidine, n-cbz-4-hydroxy-1-piperidine, benzyl 4-hydroxytetrahydro-1 2h-pyridinecarboxylate, n-benzyloxycarbonyl-4-hydroxypiperidine, 1-piperidinecarboxylic acid, 4-hydroxy-, phenylmethyl ester, n-cbz-4-hydroxypiperidine, n-benzyloxycarbonyl piperidin-4-ol, 4-hydroxypiperidine, n-cbz protected, 4-hydroxy-piperidine-1-carboxylic acid benzyl ester
C1CN(CCC1O)C(=O)OCC2=CC=CC=C2
235.283
235.28
≥98.0% (GC)
Yellow
95798-23-5
MFCD01863722
JKIUUDJOCYHIGY-UHFFFAOYSA-N
benzyl 4-hydroxypiperidine-1-carboxylate
2794620
Clear Liquid at 20°C
Benzyl 4-Hydroxy-1-piperidinecarboxylate
Safety and Handling

Safety and Handling

TSCA : No