missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl 4-bromotetrahydro-1(2H)-pyridinecarboxylate, ≥97%, Thermo Scientific™

Chemical Identifiers

CAS 166953-64-6
Molecular Formula C13H16BrNO2
Molecular Weight (g/mol) 298.18
MDL Number MFCD02681990
InChI Key BFWKSPOSMPKTSV-UHFFFAOYSA-N
Synonym 1-cbz-4-bromopiperidine, 4-bromo-n-cbz piperidine, 4-bromo-n-z-piperidine, 4-bromopiperidine, n-cbz protected, benzyl 4-bromo-1-piperidinecarboxylate, benzyl 4-bromotetrahydro-1 2h-pyridinecarboxylate, benzyl4-bromopiperidine-1-carboxylate, buttpark 9350-52, n-cbz-4-bromo-piperidine, pubchem16157
PubChem CID 2776275
IUPAC Name benzyl 4-bromopiperidine-1-carboxylate
SMILES C1CN(CCC1Br)C(=O)OCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC07510DA
SDS View Documents Promotion Details
Fisher Scientific Company
CC07510DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC07510DE
SDS View Documents Promotion Details
Fisher Scientific Company
CC07510DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

166953-64-6
298.18
BFWKSPOSMPKTSV-UHFFFAOYSA-N
2776275
C1CN(CCC1Br)C(=O)OCC2=CC=CC=C2
C13H16BrNO2
MFCD02681990
1-cbz-4-bromopiperidine, 4-bromo-n-cbz piperidine, 4-bromo-n-z-piperidine, 4-bromopiperidine, n-cbz protected, benzyl 4-bromo-1-piperidinecarboxylate, benzyl 4-bromotetrahydro-1 2h-pyridinecarboxylate, benzyl4-bromopiperidine-1-carboxylate, buttpark 9350-52, n-cbz-4-bromo-piperidine, pubchem16157
benzyl 4-bromopiperidine-1-carboxylate

Specifications

166953-64-6
100.0
97%
MFCD02681990
1g
BFWKSPOSMPKTSV-UHFFFAOYSA-N
benzyl 4-bromopiperidine-1-carboxylate
2776275
97%
96.5
1.374g/mL
C13H16BrNO2
Amber glass bottle
1-cbz-4-bromopiperidine, 4-bromo-n-cbz piperidine, 4-bromo-n-z-piperidine, 4-bromopiperidine, n-cbz protected, benzyl 4-bromo-1-piperidinecarboxylate, benzyl 4-bromotetrahydro-1 2h-pyridinecarboxylate, benzyl4-bromopiperidine-1-carboxylate, buttpark 9350-52, n-cbz-4-bromo-piperidine, pubchem16157
C1CN(CCC1Br)C(=O)OCC2=CC=CC=C2
298.18
298.18
Benzyl 4-bromotetrahydro-1(2H)-pyridinecarboxylate
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Product Certifications