missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl 4-Bromophenyl Ketone, TCI America™

$79.45 - $279.59

Chemical Identifiers

CAS 2001-29-8
Molecular Formula C14H11BrO
Molecular Weight (g/mol) 275.145
MDL Number MFCD00016331
InChI Key MOSIKPSTRPODHQ-UHFFFAOYSA-N
Synonym benzyl 4-bromophenyl ketone, 1-4-bromophenyl-2-phenylethanone, 4'-bromo-2-phenylacetophenone, 4-bromodeoxybenzoin, 4-bromodesoxybenzoin, 1-4-bromophenyl-2-phenyl-ethanone, ethanone, 1-4-bromophenyl-2-phenyl, 1-4-bromophenyl-2-phenylethan-1-one, 4-bromodeoxybenzoin, acmc-1chtj
PubChem CID 519738
IUPAC Name 1-(4-bromophenyl)-2-phenylethanone
SMILES C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B20201G View Documents TCI America
B20201G
1g Each for $79.45
 
B20205G View Documents TCI America
B20205G
5g Each for $279.59
 
Specifications

Chemical Identifiers

2001-29-8
275.145
MOSIKPSTRPODHQ-UHFFFAOYSA-N
519738
C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)Br
C14H11BrO
MFCD00016331
benzyl 4-bromophenyl ketone, 1-4-bromophenyl-2-phenylethanone, 4'-bromo-2-phenylacetophenone, 4-bromodeoxybenzoin, 4-bromodesoxybenzoin, 1-4-bromophenyl-2-phenyl-ethanone, ethanone, 1-4-bromophenyl-2-phenyl, 1-4-bromophenyl-2-phenylethan-1-one, 4-bromodeoxybenzoin, acmc-1chtj
1-(4-bromophenyl)-2-phenylethanone

Specifications

Undesignated
1g
C14H11BrO
benzyl 4-bromophenyl ketone, 1-4-bromophenyl-2-phenylethanone, 4'-bromo-2-phenylacetophenone, 4-bromodeoxybenzoin, 4-bromodesoxybenzoin, 1-4-bromophenyl-2-phenyl-ethanone, ethanone, 1-4-bromophenyl-2-phenyl, 1-4-bromophenyl-2-phenylethan-1-one, 4-bromodeoxybenzoin, acmc-1chtj
C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)Br
275.145
275.15
≥97.0% (GC)
112°C
2001-29-8
MFCD00016331
MOSIKPSTRPODHQ-UHFFFAOYSA-N
1-(4-bromophenyl)-2-phenylethanone
519738
Crystal-Powder at 20°C
Benzyl 4-Bromophenyl Ketone
Safety and Handling

Safety and Handling

TSCA : Yes