Please login to your online account to display your discounted pricing

Benzyl 4-bromophenyl ketone, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Benzyl 4-bromophenyl ketone
Assay ≥97.5%
Molecular Formula C14H11BrO
Linear Formula C6H5CH2COC6H4Br
Formula Weight 275.14
MDL Number MFCD00016331
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381890010

  SDS
thermo fisher scientific
AC381890010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC381890050

  SDS
thermo fisher scientific
AC381890050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 275.14
Assay ≥97.5%
Physical Form Crystalline Powder
Quantity 1g
Color White to pale yellow
Melting Point 110° to 115°C
Linear Formula C6H5CH2COC6H4Br
Molecular Formula C14H11BrO
CAS 2001-29-8
Chemical Name or Material Benzyl 4-bromophenyl ketone
MDL Number MFCD00016331
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.