Learn More
Benzyl 4-(bromomethyl)tetrahydro-1(2H)-pyridinecarboxylate, 97%, Maybridge™
Manufacturer: Fisher Scientific Company CC07508DA
Specifications
Benzyl 4-(bromomethyl)tetrahydro-1(2H)-pyridinecarboxylate | |
1g | |
96.5 | |
97% | |
MFCD02681989 | |
XJHKDSZGAWXUTB-UHFFFAOYSA-N | |
benzyl 4-(bromomethyl)piperidine-1-carboxylate | |
2776274 | |
97% |
Amber glass bottle | |
159275-17-9 | |
100.0 | |
C14H18BrNO2 | |
1-cbz-4-bromomethyl piperidine, 1-piperidinecarboxylic acid, 4-bromomethyl-, phenylmethyl ester, benzyl 4-bromoethyl tetrahydro-1 2h-pyridinecarboxylate, benzyl 4-bromomethyl piperidine-1-carboxylate, benzyl 4-bromomethyl tetrahydro-1 2h pyridinecarboxylate, benzyl 4-bromomethyl tetrahydro-1 2h-pyridinecarboxylate, benzyloxycarbonylbromomethylpiperidine, n-benzyloxycarbonyl-4-bromomethyl piperidine, n-cbz-4-bromomethyl piperidine, phenylmethyl 4-bromomethyl piperidinecarboxylate | |
C1CN(CCC1CBr)C(=O)OCC2=CC=CC=C2 | |
312.207 | |
312.21 |
Chemical Identifiers
159275-17-9 | |
312.207 | |
XJHKDSZGAWXUTB-UHFFFAOYSA-N | |
2776274 | |
C1CN(CCC1CBr)C(=O)OCC2=CC=CC=C2 |
C14H18BrNO2 | |
MFCD02681989 | |
1-cbz-4-bromomethyl piperidine, 1-piperidinecarboxylic acid, 4-bromomethyl-, phenylmethyl ester, benzyl 4-bromoethyl tetrahydro-1 2h-pyridinecarboxylate, benzyl 4-bromomethyl piperidine-1-carboxylate, benzyl 4-bromomethyl tetrahydro-1 2h pyridinecarboxylate, benzyl 4-bromomethyl tetrahydro-1 2h-pyridinecarboxylate, benzyloxycarbonylbromomethylpiperidine, n-benzyloxycarbonyl-4-bromomethyl piperidine, n-cbz-4-bromomethyl piperidine, phenylmethyl 4-bromomethyl piperidinecarboxylate | |
benzyl 4-(bromomethyl)piperidine-1-carboxylate |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger