Please login to your online account to display your discounted pricing

Benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material Benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate
CAS 167757-45-1
CAS Min % 94.5
CAS Max % 100.0
Molecular Formula C14H18N2O3
Formula Weight 262.31
MDL Number MFCD01568760
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

CC00094DA

  SDS
fisher scientific company
CC00094DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material Benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate
Assay 95%
Assay Percent Range 95%
Molecular Formula C14H18N2O3
Formula Weight 262.31
MDL Number MFCD01568760
CAS 167757-45-1
CAS Min % 94.5
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water fo
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water fo

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.