missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl (3-Fluoro-4-morpholinophenyl)carbamate, TCI America™

$135.97 - $458.34

Chemical Identifiers

CAS 168828-81-7
Molecular Formula C18H19FN2O3
Molecular Weight (g/mol) 330.359
MDL Number MFCD06658240
InChI Key XKGUZGHMWUIYDR-UHFFFAOYSA-N
Synonym (3-Fluoro-4-morpholinophenyl)carbamic Acid Benzyl Ester
PubChem CID 5306150
IUPAC Name benzyl N-(3-fluoro-4-morpholin-4-ylphenyl)carbamate
SMILES C1COCCN1C2=C(C=C(C=C2)NC(=O)OCC3=CC=CC=C3)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B48105G View Documents TCI America
B48105G
5g Each for $135.97
 
B481025G View Documents TCI America
B481025G
25g Each for $458.34
 
Specifications

Chemical Identifiers

168828-81-7
330.359
XKGUZGHMWUIYDR-UHFFFAOYSA-N
5306150
C1COCCN1C2=C(C=C(C=C2)NC(=O)OCC3=CC=CC=C3)F
C18H19FN2O3
MFCD06658240
(3-Fluoro-4-morpholinophenyl)carbamic Acid Benzyl Ester
benzyl N-(3-fluoro-4-morpholin-4-ylphenyl)carbamate

Specifications

White-Yellow
5g
C18H19FN2O3
(3-Fluoro-4-morpholinophenyl)carbamic Acid Benzyl Ester
C1COCCN1C2=C(C=C(C=C2)NC(=O)OCC3=CC=CC=C3)F
330.359
330.36
≥98.0% (HPLC,N)
125°C
168828-81-7
MFCD06658240
XKGUZGHMWUIYDR-UHFFFAOYSA-N
benzyl N-(3-fluoro-4-morpholin-4-ylphenyl)carbamate
5306150
Crystal-Powder at 20°C
Benzyl (3-Fluoro-4-morpholinophenyl)carbamate
Safety and Handling

Safety and Handling

TSCA : No