Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl 2-naphthyl ether, 99%

$81.22 - $578.59

Chemical Identifiers

Chemical Name or Material Benzyl 2-naphthyl ether
CAS 613-62-7
Molecular Formula C17H14O
Formula Weight 234.3
MDL Number MFCD00089092
Beilstein 2049922
Synonym 2-Benzyloxynaphthalene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2179614

  SDS
alfa aesar™
B2179614
25g Each for $81.22

AAB2179622

  SDS
alfa aesar™
B2179622
100g Each for $215.47

AAB2179636

  SDS
alfa aesar™
B2179636
500g Each for $578.59
Description & Specifications

Specifications

Formula Weight 234.3
Synonym 2-Benzyloxynaphthalene
Chemical Name or Material Benzyl 2-naphthyl ether
Molecular Formula C17H14O
Assay 99%
Beilstein 2049922
Melting Point 96° to 98°C
MDL Number MFCD00089092
CAS 613-62-7
Quantity 25g