missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyl 2-(hydroxymethyl)-1-indolinecarboxylate, ≥95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC13409DE
Specifications
Benzyl 2-(hydroxymethyl)-1-indolinecarboxylate | |
94.5 | |
95% | |
MFCD02682013 | |
benzyl 2-hydroxymethyl-1-indolinecarboxylate, benzyl 2-hydroxymethyl indoline-1-carboxylate, benzyl 2-hydroxymethyl-2,3-dihydroindole-1-carboxylate, 1h-indole-1-carboxylicacid, 2,3-dihydro-2-hydroxymethyl-, phenylmethyl ester, acmc-20c3wg, 1-benzyloxycarbonylindoline-2-methanol, +/-1-benzyloxycarbonyl-2-hydroxymethylindoline | |
C1C(N(C2=CC=CC=C21)C(=O)OCC3=CC=CC=C3)CO | |
283.327 | |
283.33 |
135829-04-8 | |
100.0 | |
C17H17NO3 | |
5g | |
QSMOQTIGILELKY-UHFFFAOYSA-N | |
benzyl 2-(hydroxymethyl)-2,3-dihydroindole-1-carboxylate | |
2776402 | |
95% |
Chemical Identifiers
135829-04-8 | |
283.327 | |
QSMOQTIGILELKY-UHFFFAOYSA-N | |
2776402 | |
C1C(N(C2=CC=CC=C21)C(=O)OCC3=CC=CC=C3)CO |
C17H17NO3 | |
MFCD02682013 | |
benzyl 2-hydroxymethyl-1-indolinecarboxylate, benzyl 2-hydroxymethyl indoline-1-carboxylate, benzyl 2-hydroxymethyl-2,3-dihydroindole-1-carboxylate, 1h-indole-1-carboxylicacid, 2,3-dihydro-2-hydroxymethyl-, phenylmethyl ester, acmc-20c3wg, 1-benzyloxycarbonylindoline-2-methanol, +/-1-benzyloxycarbonyl-2-hydroxymethylindoline | |
benzyl 2-(hydroxymethyl)-2,3-dihydroindole-1-carboxylate |