missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl 2-bromoethyl ether, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC388870010

 View more versions of this product

Catalog No. AC388870010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

215.09
1.36g/mL
100.0
1462-37-9
97%
Glass bottle
1g
96.0
C6H5CH2OCH2CH2Br
Benzyl 2-bromoethyl ether
MFCD01321307
C9H11BrO

Chemical Identifiers

1462-37-9
100.0
C9H11BrO
215.09
Benzyl 2-bromoethyl ether
96.0
97%
C6H5CH2OCH2CH2Br
MFCD01321307

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning