Learn More
Benzyl 2-bromoethyl ether, 97%, Thermo Scientific Chemicals
$51.73 - $748.34
Chemical Identifiers
CAS | 1462-37-9 |
---|---|
Molecular Formula | C9H11BrO |
Molecular Weight (g/mol) | 215.09 |
MDL Number | MFCD01321307 |
InChI Key | FWOHDAGPWDEWIB-UHFFFAOYSA-N |
Synonym | benzyl 2-bromoethyl ether, 2-bromoethoxy methyl benzene, 2-benzyloxy-1-bromoethane, benzyloxyethyl bromide, benzene, 2-bromoethoxy methyl, 1-bromo-2-benzyloxyethane, 2-bromoethoxymethyl benzene, 2-bromo-ethoxymethyl-benzene |
PubChem CID | 73833 |
IUPAC Name | 2-bromoethoxymethylbenzene |
SMILES | C1=CC=C(C=C1)COCCBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5687203
|
Thermo Scientific Chemicals
H5687203 |
1 g |
Each for $51.73
|
|
|||||
AAH5687206
|
Thermo Scientific Chemicals
H5687206 |
5 g |
Each for $202.92
|
|
|||||
AAH5687214
|
Thermo Scientific Chemicals
H5687214 |
25 g |
Each for $748.34
|
|
|||||
Description
Alkylation reagent for synthesis of a macrocyclic antifungal antibiotic. Building block used to prepare a copper phthalocyanine discotic liquid crystal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAlkylation reagent for synthesis of a macrocyclic antifungal antibiotic. Building block used to prepare a copper phthalocyanine discotic liquid crystal.
Solubility
Slightly soluble in water
Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
Chemical Identifiers
1462-37-9 | |
215.09 | |
FWOHDAGPWDEWIB-UHFFFAOYSA-N | |
73833 | |
C1=CC=C(C=C1)COCCBr |
C9H11BrO | |
MFCD01321307 | |
benzyl 2-bromoethyl ether, 2-bromoethoxy methyl benzene, 2-benzyloxy-1-bromoethane, benzyloxyethyl bromide, benzene, 2-bromoethoxy methyl, 1-bromo-2-benzyloxyethane, 2-bromoethoxymethyl benzene, 2-bromo-ethoxymethyl-benzene | |
2-bromoethoxymethylbenzene |
Specifications
1462-37-9 | |
254°C | |
C9H11BrO | |
C6H5CH2OCH2CH2Br | |
1 g | |
Slightly soluble in water | |
C1=CC=C(C=C1)COCCBr | |
215.09 | |
215.09 | |
Benzyl 2-bromoethyl ether |
1.36 | |
>113°C (235°F) | |
1.541 | |
MFCD01321307 | |
benzyl 2-bromoethyl ether, 2-bromoethoxy methyl benzene, 2-benzyloxy-1-bromoethane, benzyloxyethyl bromide, benzene, 2-bromoethoxy methyl, 1-bromo-2-benzyloxyethane, 2-bromoethoxymethyl benzene, 2-bromo-ethoxymethyl-benzene | |
FWOHDAGPWDEWIB-UHFFFAOYSA-N | |
2-bromoethoxymethylbenzene | |
73833 | |
97% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only