missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl 2-acetamido-4,6-O-benzylidene-2-deoxy-alpha-D-glucopyranoside

$96.84 - $704.74

Chemical Identifiers

CAS 13343-63-0
Molecular Formula C22H25NO6
Molecular Weight (g/mol) 399.443
MDL Number MFCD00143679
InChI Key NXGXFAKJUWEFEC-NVZUTRPHSA-N
Synonym n-4ar,6s,7r,8r,8as-6-benzyloxy-8-hydroxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-7-yl acetamide, benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-a-d-glcopyranoside, benzyl-2-acetamido-4,6-o-benzylidene-2-deoxy-, a-d-glucopyranoside, benzyl-2-acetamido-4,6-o-benzylidene-2-deoxy-alpha-d-glcopyranoside, benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-alpha-d-glucopyranoside, n-4ar,6s,7r,8r,8as-6-benzyloxy-8-hydroxy-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxin-7-yl acetamide, maybridge1_006454, benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-, a-d-glucopyranoside, phenylmethyl 2-acetylamino-2-deoxy-4,6-o-phenylmethylene-, a-d-glucopyranoside, 1-o-benzyl-n-acetyl-4-o,6-o-benzylidene-alpha-d-glucosamine
PubChem CID 2735220
IUPAC Name N-[(4aR,6S,7R,8R,8aS)-8-hydroxy-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl]acetamide
SMILES CC(=O)NC1C(C2C(COC(O2)C3=CC=CC=C3)OC1OCC4=CC=CC=C4)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66981ME View Documents Alfa Aesar™
J66981ME
500mg Each for $96.84
 
AAJ6698103 View Documents Alfa Aesar™
J6698103
1g Each for $175.80
 
AAJ6698106 View Documents Alfa Aesar™
J6698106
5g Each for $704.74
 
Specifications

Chemical Identifiers

13343-63-0
399.443
NXGXFAKJUWEFEC-NVZUTRPHSA-N
2735220
CC(=O)NC1C(C2C(COC(O2)C3=CC=CC=C3)OC1OCC4=CC=CC=C4)O
C22H25NO6
MFCD00143679
n-4ar,6s,7r,8r,8as-6-benzyloxy-8-hydroxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-7-yl acetamide, benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-a-d-glcopyranoside, benzyl-2-acetamido-4,6-o-benzylidene-2-deoxy-, a-d-glucopyranoside, benzyl-2-acetamido-4,6-o-benzylidene-2-deoxy-alpha-d-glcopyranoside, benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-alpha-d-glucopyranoside, n-4ar,6s,7r,8r,8as-6-benzyloxy-8-hydroxy-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxin-7-yl acetamide, maybridge1_006454, benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-, a-d-glucopyranoside, phenylmethyl 2-acetylamino-2-deoxy-4,6-o-phenylmethylene-, a-d-glucopyranoside, 1-o-benzyl-n-acetyl-4-o,6-o-benzylidene-alpha-d-glucosamine
N-[(4aR,6S,7R,8R,8aS)-8-hydroxy-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl]acetamide

Specifications

White
500mg
C22H25NO6
55951
NXGXFAKJUWEFEC-NVZUTRPHSA-N
N-[(4aR,6S,7R,8R,8aS)-8-hydroxy-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl]acetamide
2735220
Crystalline
256°C to 261°C
13343-63-0
MFCD00143679
n-4ar,6s,7r,8r,8as-6-benzyloxy-8-hydroxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-7-yl acetamide, benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-a-d-glcopyranoside, benzyl-2-acetamido-4,6-o-benzylidene-2-deoxy-, a-d-glucopyranoside, benzyl-2-acetamido-4,6-o-benzylidene-2-deoxy-alpha-d-glcopyranoside, benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-alpha-d-glucopyranoside, n-4ar,6s,7r,8r,8as-6-benzyloxy-8-hydroxy-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxin-7-yl acetamide, maybridge1_006454, benzyl 2-acetamido-4,6-o-benzylidene-2-deoxy-, a-d-glucopyranoside, phenylmethyl 2-acetylamino-2-deoxy-4,6-o-phenylmethylene-, a-d-glucopyranoside, 1-o-benzyl-n-acetyl-4-o,6-o-benzylidene-alpha-d-glucosamine
CC(=O)NC1C(C2C(COC(O2)C3=CC=CC=C3)OC1OCC4=CC=CC=C4)O
399.443
399.44
Benzyl 2-acetamido-4,6-O-benzylidene-2-deoxy-alpha-D-glucopyranoside