Please login to your online account to display your discounted pricing

Benzyl 2,2,2-trichloroacetimidate, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  351600250

 View more versions of this product

Catalog No. AC351600250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 252.53
Physical Form Liquid
Quantity 25g
Color Clear colorless to yellow
CAS Max % 100.0
Refractive Index 1.5435 to 1.5455 (20°C, 589nm)
Chemical Name or Material Benzyl 2, 2, 2-trichloroacetimidate
Packaging Glass bottle
Linear Formula CCl3C(=NH)OCHC6H5
Molecular Formula C9H8Cl3NO
CAS 81927-55-1
Assay Percent Range 99%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
MDL Number MFCD00000805
CAS Min % 98.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Benzyl 2, 2, 2-trichloroacetimidate
CAS 81927-55-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H8Cl3NO
Linear Formula CCl3C(=NH)OCHC6H5
Formula Weight 252.53
MDL Number MFCD00000805
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents