missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl 2,2,2-trichloroacetimidate, 98%

Manufacturer:  Alfa Aesar™ A1238714

 View more versions of this product

Catalog No. AAA1238714


 

Description

Description

Applications
Benzyl 2,2,2-trichloroacetimidate is used for the acid-catalyzed benzylation of hydroxy groups. It acts as reagent during the synthesis of funiculosin dimethyl ether and (S)-3-(benzyloxy)-2-methylpropanal. It can also be used for mild esterification of carboxylic acids in the presence of a catalytic amount of BF3 etherate.

Solubility
Miscible with cyclohexane/dichloromethane.

Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

Benzyl 2,2,2-trichloroacetimidate
98%
MFCD00000805
2,2,2-Trichloroacetimidic acid benzyl ester
HUZCTWYDQIQZPM-UHFFFAOYSA-N
benzyl 2,2,2-trichloroethanimidate
25g
144968
81927-55-1
C9H8Cl3NO
2525375
110° to 114°C (0.5mmHg)
C1=CC=C(C=C1)COC(=N)C(Cl)(Cl)Cl
252.519
>110°C (230°F)
252.53

Chemical Identifiers

81927-55-1
252.519
HUZCTWYDQIQZPM-UHFFFAOYSA-N
144968
C9H8Cl3NO
MFCD00000805
2,2,2-Trichloroacetimidic acid benzyl ester
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2°C to 8°C

SDS Documents