missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzydamine hydrochloride, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6118014

 View more versions of this product

Catalog No. AAJ6118014



Description

Description

Nonsteroidal analgesic and anti-inflammatory compound. A prostaglandin synthase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Nonsteroidal analgesic and anti-inflammatory compound
  • A prostaglandin synthase inhibitor
Specifications

Specifications

Benzydamine hydrochloride
156°C to 161°C
C19H24ClN3O
25 g
benzydamine hydrochloride, benzydamine hcl, benalgin, benzindamine hydrochloride, benzidamine hydrochloride, benzidan, benzyrin, difflam, dorinamin, enzamin
HNNIWKQLJSNAEQ-UHFFFAOYSA-N
3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride
65464
Crystalline Powder
132-69-4
White
MFCD00078957
14,1122
Soluble in water,ethanol,chloroform,n-butanol or DMSO /n
CN(C)CCCOC1=NN(C2=CC=CC=C21)CC3=CC=CC=C3.Cl
345.871
345.87

Chemical Identifiers

132-69-4
345.871
HNNIWKQLJSNAEQ-UHFFFAOYSA-N
65464
CN(C)CCCOC1=NN(C2=CC=CC=C21)CC3=CC=CC=C3.Cl
C19H24ClN3O
MFCD00078957
benzydamine hydrochloride, benzydamine hcl, benalgin, benzindamine hydrochloride, benzidamine hydrochloride, benzidan, benzyrin, difflam, dorinamin, enzamin
3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P363-P501c

H302+H312+H332-H319

EINECSNumber : 205-076-0

RTECSNumber : NK7875000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only