Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzoylurea, 97%

$34.32 - $34.32

Chemical Identifiers

Chemical Name or Material Benzoylurea
CAS 614-22-2
Molecular Formula C6H5CONHCONH2
Formula Weight 164.17
MDL Number MFCD00025441
Beilstein 2045538
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1131306

 
alfa aesar™
L1131306
5g Each for $34.32
Description & Specifications

Specifications

Formula Weight 164.17
Assay 97%
CAS 614-22-2
Chemical Name or Material Benzoylurea
Molecular Formula C6H5CONHCONH2
MDL Number MFCD00025441
Beilstein 2045538
Melting Point 209° to 210°C
Quantity 5g
RTECS Number YR9070000