missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzoylurea, 97%

Manufacturer:  Alfa Aesar™ L1131314

 View more versions of this product

Catalog No. AAL1131314



Specifications

Specifications

Benzoylurea
25g
C8H8N2O2
2045538
HRYILSDLIGTCOP-UHFFFAOYSA-N
N-carbamoylbenzamide
69190
164.17
209°C to 210°C
614-22-2
MFCD00025441
benzoylurea, 1-benzoylurea, n-benzoylurea, urea, benzoyl, benzoic acid, ureide, n-aminocarbonyl benzamide, benzamide, n-aminocarbonyl, benzamide, n-aminocarbonyl-9ci, benzoyl urea, benzoylcarbamide
C1=CC=C(C=C1)C(=O)NC(=O)N
164.164
CHEBI:39416
97%

Chemical Identifiers

614-22-2
164.164
HRYILSDLIGTCOP-UHFFFAOYSA-N
69190
N-carbamoylbenzamide
C8H8N2O2
MFCD00025441
benzoylurea, 1-benzoylurea, n-benzoylurea, urea, benzoyl, benzoic acid, ureide, n-aminocarbonyl benzamide, benzamide, n-aminocarbonyl, benzamide, n-aminocarbonyl-9ci, benzoyl urea, benzoylcarbamide
CHEBI:39416
C1=CC=C(C=C1)C(=O)NC(=O)N
Safety and Handling

Safety and Handling

RTECSNumber : YR9070000

Documents