Learn More
Benzoylnitromethane, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 614-21-1 |
---|---|
Molecular Formula | C8H7NO3 |
Molecular Weight (g/mol) | 165.15 |
MDL Number | MFCD00010218 |
InChI Key | JTWHVBNYYWFXSI-UHFFFAOYSA-N |
Synonym | benzoylnitromethane, ethanone, 2-nitro-1-phenyl, alpha-nitroacetophenone, 2-nitro-1-phenylethanon, nitroacetophenone, alpha-nitro acetophenone, acmc-1bdpo, acetophenone, 2-nitro, nitromethyl phenyl ketone, .alpha.-nitroacetophenone |
PubChem CID | 94833 |
IUPAC Name | 2-nitro-1-phenylethanone |
SMILES | C1=CC=C(C=C1)C(=O)C[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC372790050
|
Thermo Scientific Chemicals
372790050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC372790250
|
Thermo Scientific Chemicals
372790250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
614-21-1 | |
165.15 | |
JTWHVBNYYWFXSI-UHFFFAOYSA-N | |
94833 | |
C1=CC=C(C=C1)C(=O)C[N+](=O)[O-] |
C8H7NO3 | |
MFCD00010218 | |
benzoylnitromethane, ethanone, 2-nitro-1-phenyl, alpha-nitroacetophenone, 2-nitro-1-phenylethanon, nitroacetophenone, alpha-nitro acetophenone, acmc-1bdpo, acetophenone, 2-nitro, nitromethyl phenyl ketone, .alpha.-nitroacetophenone | |
2-nitro-1-phenylethanone |
Specifications
614-21-1 | |
100.0 | |
97.5% min. (GC) | |
C8H7NO3 | |
MFCD00010218 | |
benzoylnitromethane, ethanone, 2-nitro-1-phenyl, alpha-nitroacetophenone, 2-nitro-1-phenylethanon, nitroacetophenone, alpha-nitro acetophenone, acmc-1bdpo, acetophenone, 2-nitro, nitromethyl phenyl ketone, .alpha.-nitroacetophenone | |
JTWHVBNYYWFXSI-UHFFFAOYSA-N | |
2-nitro-1-phenylethanone | |
94833 | |
98% |
97.5 | |
Authentic | |
Glass bottle | |
C6H5C(O)CH2NO2 | |
5g | |
Solubility in water: .. Other solubilities: soluble in acetone,halogenated solvents,thf and,alcohols | |
C1=CC=C(C=C1)C(=O)C[N+](=O)[O-] | |
165.15 | |
165.15 | |
Benzoylnitromethane |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
RUO â Research Use Only