Learn More
Benzoylnitromethane, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 372790250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Benzoylnitromethane | |
97.5 | |
Authentic | |
Glass bottle | |
C6H5C(O)CH2NO2 | |
25g | |
Solubility in water: .. Other solubilities: soluble in acetone,halogenated solvents,thf and,alcohols | |
C1=CC=C(C=C1)C(=O)C[N+](=O)[O-] | |
165.15 | |
165.15 |
614-21-1 | |
100.0 | |
97.5% min. (GC) | |
C8H7NO3 | |
MFCD00010218 | |
benzoylnitromethane, ethanone, 2-nitro-1-phenyl, alpha-nitroacetophenone, 2-nitro-1-phenylethanon, nitroacetophenone, alpha-nitro acetophenone, acmc-1bdpo, acetophenone, 2-nitro, nitromethyl phenyl ketone, .alpha.-nitroacetophenone | |
JTWHVBNYYWFXSI-UHFFFAOYSA-N | |
2-nitro-1-phenylethanone | |
94833 | |
98% |
Chemical Identifiers
614-21-1 | |
165.15 | |
JTWHVBNYYWFXSI-UHFFFAOYSA-N | |
94833 | |
C1=CC=C(C=C1)C(=O)C[N+](=O)[O-] |
C8H7NO3 | |
MFCD00010218 | |
benzoylnitromethane, ethanone, 2-nitro-1-phenyl, alpha-nitroacetophenone, 2-nitro-1-phenylethanon, nitroacetophenone, alpha-nitro acetophenone, acmc-1bdpo, acetophenone, 2-nitro, nitromethyl phenyl ketone, .alpha.-nitroacetophenone | |
2-nitro-1-phenylethanone |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
RUO â Research Use Only